gRNA Design Details
- Target DNA Sequence
- AACTGTCCAAAAAGAACCGG
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 121827308 - 121827330 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
87.21/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ccdc13
- Entrez Gene ID
-
100502861
- Gene Name
- coiled-coil domain containing 13
- Gene Aliases
-
2900041A11Rik, Gm188
- Chromosome Location
-
Chr.9: 121797624 - 121839515 on GRCm38
- UniGene ID
- Mm.335550
- Species
-
Mouse