gRNA Design Details
- Target DNA Sequence
- AACCGTAAGAGCTTGGTCGT
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 70790050 - 70790072 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mast3
- Entrez Gene ID
-
546071
- Gene Name
- microtubule associated serine/threonine kinase 3
- Gene Aliases
-
BC024265, mKIAA0561
- Chromosome Location
-
Chr.8: 70778117 - 70805068 on GRCm38
- UniGene ID
- Mm.250731
- Species
-
Mouse