gRNA Design Details
- Target DNA Sequence
- TCCAGCTCCGAATCCAAAGG
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 83693961 - 83693983 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pkn1
- Entrez Gene ID
-
320795
- Gene Name
- protein kinase N1
- Gene Aliases
-
DBK, F730027O18Rik, PAK1, PRK1, Pkn, Prkcl1, Stk3
- Chromosome Location
-
Chr.8: 83669762 - 83699179 on GRCm38
- UniGene ID
- Mm.213000
- Species
-
Mouse