gRNA Design Details
- Target DNA Sequence
- AGCGACTGAAGCGGGAGATA
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 83692817 - 83692839 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.51/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pkn1
- Entrez Gene ID
-
320795
- Gene Name
- protein kinase N1
- Gene Aliases
-
DBK, F730027O18Rik, PAK1, PRK1, Pkn, Prkcl1, Stk3
- Chromosome Location
-
Chr.8: 83669762 - 83699179 on GRCm38
- UniGene ID
- Mm.213000
- Species
-
Mouse