gRNA Design Details
- Target DNA Sequence
- TCCCATTTGGGTTGATGCGC
- PAM Sequence
- CGG
- Target locus
-
Chr.8: 83918838 - 83918860 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrl1
- Entrez Gene ID
-
330814
- Gene Name
- adhesion G protein-coupled receptor L1
- Gene Aliases
-
2900070I05Rik, AI182398, CLIBA, Lec2, Lphn1, mKIAA0821
- Chromosome Location
-
Chr.8: 83900098 - 83941954 on GRCm38
- UniGene ID
- Mm.260733
- Species
-
Mouse