gRNA Design Details
- Target DNA Sequence
- TAACCGAACCCAGTGTGTGG
- PAM Sequence
- TGG
- Target locus
-
Chr.8: 83923005 - 83923027 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrl1
- Entrez Gene ID
-
330814
- Gene Name
- adhesion G protein-coupled receptor L1
- Gene Aliases
-
2900070I05Rik, AI182398, CLIBA, Lec2, Lphn1, mKIAA0821
- Chromosome Location
-
Chr.8: 83900098 - 83941954 on GRCm38
- UniGene ID
- Mm.260733
- Species
-
Mouse