gRNA Design Details
- Target DNA Sequence
- GGGTACACTTGAACCAGTTG
- PAM Sequence
- AGG
- Target locus
-
Chr.9: 64506100 - 64506122 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
90.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Megf11
- Entrez Gene ID
-
214058
- Gene Name
- multiple EGF-like-domains 11
- Gene Aliases
-
2410080H04Rik, D130061K05
- Chromosome Location
-
Chr.9: 64385618 - 64709205 on GRCm38
- UniGene ID
- Mm.127721
- Species
-
Mouse