gRNA Design Details
- Target DNA Sequence
- AAAGCTGCCTCGCCCAATTA
- PAM Sequence
- GGG
- Target locus
-
Chr.8: 105912994 - 105913016 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
92.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pskh1
- Entrez Gene ID
-
244631
- Gene Name
- protein serine kinase H1
- Gene Aliases
-
AW539964, E130013P03Rik, b2b1230Clo
- Chromosome Location
-
Chr.8: 105900474 - 105931802 on GRCm38
- UniGene ID
- Mm.27627
- Species
-
Mouse