gRNA Design Details
- Target DNA Sequence
- CTCAGCGACTTGAGCAGAGG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 6113942 - 6113964 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.2/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Proser2
- Entrez Gene ID
-
227545
- Gene Name
- proline and serine rich 2
- Gene Aliases
-
5430407P10Rik, AU067770
- Chromosome Location
-
Chr.2: 6098493 - 6130294 on GRCm38
- UniGene ID
- Mm.133542
- Species
-
Mouse