gRNA Design Details

Target DNA Sequence
CTCAGCGACTTGAGCAGAGG
PAM Sequence
TGG
Target locus
Chr.2: 6113942 - 6113964 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 88.2/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Proser2
Entrez Gene ID
227545
Gene Name
proline and serine rich 2
Gene Aliases
5430407P10Rik, AU067770
Chromosome Location
Chr.2: 6098493 - 6130294 on GRCm38
UniGene ID
Mm.133542
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001159657.1 NP_001153129.1 2
NM_144883.4 NP_659132.2 2