gRNA Design Details
- Target DNA Sequence
- CTCGTCAAAGTCATATTCCA
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 6102970 - 6102992 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Proser2
- Entrez Gene ID
-
227545
- Gene Name
- proline and serine rich 2
- Gene Aliases
-
5430407P10Rik, AU067770
- Chromosome Location
-
Chr.2: 6098493 - 6130294 on GRCm38
- UniGene ID
- Mm.133542
- Species
-
Mouse