gRNA Design Details

Target DNA Sequence
CTCCTCCGAAGGAGTTTGGG
PAM Sequence
TGG
Target locus
Chr.15: 101135749 - 101135771 on GRCm38
Strand
Reverse
Application
Gene Knockout
Score: 89.46/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Acvrl1
Entrez Gene ID
11482
Gene Name
activin A receptor, type II-like 1
Gene Aliases
AI115505, AI427544, Acvrlk1, Alk1
Chromosome Location
Chr.15: 101128522 - 101145336 on GRCm38
UniGene ID
Mm.279542
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001277255.1 NP_001264184.1 6
NM_001277257.1 NP_001264186.1 5
NM_001277258.1 NP_001264187.1 5
NM_001277259.1 NP_001264188.1 6
NM_009612.3 NP_033742.2 5