gRNA Design Details

Target DNA Sequence
CAACGTGTCTCTGATGCTGG
PAM Sequence
AGG
Target locus
Chr.15: 101135521 - 101135543 on GRCm38
Strand
Forward
Application
Gene Knockout
Score: 86.31/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Acvrl1
Entrez Gene ID
11482
Gene Name
activin A receptor, type II-like 1
Gene Aliases
AI115505, AI427544, Acvrlk1, Alk1
Chromosome Location
Chr.15: 101128522 - 101145336 on GRCm38
UniGene ID
Mm.279542
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001277255.1 NP_001264184.1 5
NM_001277257.1 NP_001264186.1 4
NM_001277258.1 NP_001264187.1 4
NM_001277259.1 NP_001264188.1 5
NM_009612.3 NP_033742.2 4