gRNA Design Details
- Target DNA Sequence
- CGTCGTGGTCTGGTTAGTCT
- PAM Sequence
- GGG
- Target locus
-
Chr.9: 110858328 - 110858350 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
91.82/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss50
- Entrez Gene ID
-
235631
- Gene Name
- protease, serine 50
- Gene Aliases
-
79H19C, Tsp50
- Chromosome Location
-
Chr.9: 110857899 - 110864629 on GRCm38
- UniGene ID
- Mm.102265
- Species
-
Mouse