gRNA Design Details
- Target DNA Sequence
- TGGCTCGGAAACTCTGTCGG
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 103334042 - 103334064 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
93.08/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Mast4
- Entrez Gene ID
-
328329
- Gene Name
- microtubule associated serine/threonine kinase family member 4
- Gene Aliases
-
4930420O11Rik, AI642422, mKIAA0303
- Chromosome Location
-
Chr.13: 102732488 - 103335840 on GRCm38
- UniGene ID
- Mm.202606
- Species
-
Mouse