gRNA Design Details
- Target DNA Sequence
- GGTGGAAATTCTGTGTTCCG
- PAM Sequence
- GGG
- Target locus
-
Chr.2: 58477784 - 58477806 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
89.29/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Acvr1
- Entrez Gene ID
-
11477
- Gene Name
- activin A receptor, type 1
- Gene Aliases
-
ALK2, ActR-I, ActRIA, Acvr, Acvrlk2, Alk-2, Alk8, D330013D15Rik, SKR1, Tsk7L
- Chromosome Location
-
Chr.2: 58446438 - 58566828 on GRCm38
- UniGene ID
- Mm.689
- Species
-
Mouse