gRNA Design Details
- Target DNA Sequence
- CCGAAAACGCTTACTGGCCT
- PAM Sequence
- TGG
- Target locus
-
Chr.5: 30203498 - 30203520 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrf3
- Entrez Gene ID
-
381628
- Gene Name
- adhesion G protein-coupled receptor F3
- Gene Aliases
-
Gm1041, Gpr113, Pgr23
- Chromosome Location
-
Chr.5: 30193431 - 30205754 on GRCm38
- UniGene ID
- Mm.291148
- Species
-
Mouse