gRNA Design Details
- Target DNA Sequence
- ATTTGCAACGACGGCTATGA
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 84585512 - 84585534 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Dner
- Entrez Gene ID
-
227325
- Gene Name
- delta/notch-like EGF-related receptor
- Gene Aliases
-
A930026D19Rik, BET, Bret
- Chromosome Location
-
Chr.1: 84369839 - 84696221 on GRCm38
- UniGene ID
- Mm.292560
- Species
-
Mouse