gRNA Design Details
- Target DNA Sequence
- AGCCGTCGTTGCAAATGCAG
- PAM Sequence
- AGG
- Target locus
-
Chr.1: 84585519 - 84585541 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Dner
- Entrez Gene ID
-
227325
- Gene Name
- delta/notch-like EGF-related receptor
- Gene Aliases
-
A930026D19Rik, BET, Bret
- Chromosome Location
-
Chr.1: 84369839 - 84696221 on GRCm38
- UniGene ID
- Mm.292560
- Species
-
Mouse