gRNA Design Details
- Target DNA Sequence
- CGGTGTAACAGGATTTGAAG
- PAM Sequence
- TGG
- Target locus
-
Chr.2: 48873319 - 48873341 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.03/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Acvr2a
- Entrez Gene ID
-
11480
- Gene Name
- activin receptor IIA
- Gene Aliases
-
ActrIIa, Acvr2, TactrII
- Chromosome Location
-
Chr.2: 48814109 - 48903264 on GRCm38
- UniGene ID
- Mm.314338
- Species
-
Mouse