gRNA Design Details
- Target DNA Sequence
- TTCGGACATATCCATATACC
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 24044263 - 24044285 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
90.12/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss27
- Entrez Gene ID
-
213171
- Gene Name
- protease, serine 27
- Gene Aliases
-
CAPH2, Mpn, marapsin
- Chromosome Location
-
Chr.17: 24038243 - 24048378 on GRCm38
- UniGene ID
- Mm.133083
- Species
-
Mouse