gRNA Design Details
- Target DNA Sequence
- ATGTTCAACCGGATGGTGGG
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 24042819 - 24042841 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
88.54/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss27
- Entrez Gene ID
-
213171
- Gene Name
- protease, serine 27
- Gene Aliases
-
CAPH2, Mpn, marapsin
- Chromosome Location
-
Chr.17: 24038243 - 24048378 on GRCm38
- UniGene ID
- Mm.133083
- Species
-
Mouse