gRNA Design Details
- Target DNA Sequence
- AGGCGTAGCCGGACTTGAGT
- PAM Sequence
- AGG
- Target locus
-
Chr.16: 22162840 - 22162862 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
85.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igf2bp2
- Entrez Gene ID
-
319765
- Gene Name
- insulin-like growth factor 2 mRNA binding protein 2
- Gene Aliases
-
C330012H03Rik, IMP-2, Imp2, Neilsen
- Chromosome Location
-
Chr.16: 22059009 - 22163299 on GRCm38
- UniGene ID
- Mm.489442
- Species
-
Mouse