gRNA Design Details
- Target DNA Sequence
- TGTTCTGCGGTTTCATGTCG
- PAM Sequence
- CGG
- Target locus
-
Chr.1: 74605454 - 74605476 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Stk36
- Entrez Gene ID
-
269209
- Gene Name
- serine/threonine kinase 36
- Gene Aliases
-
1700112N14Rik, B930045J24, FU, Fused, mKIAA1278
- Chromosome Location
-
Chr.1: 74601450 - 74636894 on GRCm38
- UniGene ID
- Mm.310974
- Species
-
Mouse