gRNA Design Details
- Target DNA Sequence
- TCCCCGTTTGACCCACAGAC
- PAM Sequence
- AGG
- Target locus
-
Chr.4: 48610431 - 48610453 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
88.78/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Tmeff1
- Entrez Gene ID
-
230157
- Gene Name
- transmembrane protein with EGF-like and two follistatin-like domains 1
- Gene Aliases
-
A830033E11, M7365, Tr1
- Chromosome Location
-
Chr.4: 48585114 - 48663131 on GRCm38
- UniGene ID
- Mm.422686
- Species
-
Mouse