gRNA Design Details
- Target DNA Sequence
- CTTGAGGTCACACCTGGTGC
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 151925410 - 151925432 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
83.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Angpt4
- Entrez Gene ID
-
11602
- Gene Name
- angiopoietin 4
- Gene Aliases
-
Agpt4, Ang3
- Chromosome Location
-
Chr.2: 151911332 - 151944654 on GRCm38
- UniGene ID
- Mm.189536
- Species
-
Mouse