gRNA Design Details
- Target DNA Sequence
- CGGAAGCGGAAACTTTTCCC
- PAM Sequence
- AGG
- Target locus
-
Chr.13: 119727610 - 119727632 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
89.7/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Nim1k
- Entrez Gene ID
-
245269
- Gene Name
- NIM1 serine/threonine protein kinase
- Gene Aliases
-
E130304F04Rik, Nim1
- Chromosome Location
-
Chr.13: 119710094 - 119755882 on GRCm38
- UniGene ID
- Mm.403943
- Species
-
Mouse