gRNA Design Details

Target DNA Sequence
TCCATGACGTACACACGTTC
PAM Sequence
AGG
Target locus
Chr.18: 21168758 - 21168780 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 87.59/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Garem
Entrez Gene ID
381126
Gene Name
GRB2 associated, regulator of MAPK1
Gene Aliases
C86169, Fam59a, Gm944, mKIAA4238
Chromosome Location
Chr.18: 21127342 - 21300139 on GRCm38
UniGene ID
Mm.312276
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001033445.2 NP_001028617.2 4