gRNA Design Details
- Target DNA Sequence
- GGTTGAACAATGGGCTCTGT
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 43404017 - 43404039 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.37/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrf5
- Entrez Gene ID
-
224792
- Gene Name
- adhesion G protein-coupled receptor F5
- Gene Aliases
-
8430401C09Rik, 9330185D23, AI528491, Gpr116, mKIAA0758
- Chromosome Location
-
Chr.17: 43360329 - 43459557 on GRCm38
- UniGene ID
- Mm.23478
- Species
-
Mouse