gRNA Design Details
- Target DNA Sequence
- TTCCATCAACGTATCCATTA
- PAM Sequence
- TGG
- Target locus
-
Chr.17: 42713210 - 42713232 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
82.49/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adgrf2
- Entrez Gene ID
-
435529
- Gene Name
- adhesion G protein-coupled receptor F2
- Gene Aliases
-
Gpr111, PGR20
- Chromosome Location
-
Chr.17: 42694028 - 42749145 on GRCm38
- UniGene ID
- Mm.136350
- Species
-
Mouse