gRNA Design Details
- Target DNA Sequence
- TGTGAGTGGTTCCGCTATGG
- PAM Sequence
- AGG
- Target locus
-
Chr.2: 164569888 - 164569910 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
87.27/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Spint3
- Entrez Gene ID
-
629747
- Gene Name
- serine peptidase inhibitor, Kunitz type, 3
- Gene Aliases
-
Gm14315, OTTMUSG00000016288
- Chromosome Location
-
Chr.2: 164569695 - 164573456 on GRCm38
- UniGene ID
- Mm.234248
- Species
-
Mouse