gRNA Design Details
- Target DNA Sequence
- CCTCATTGATCGAAGCTGGG
- PAM Sequence
- TGG
- Target locus
-
Chr.9: 110838521 - 110838543 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
89.36/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss45
- Entrez Gene ID
-
260408
- Gene Name
- protease, serine 45
- Gene Aliases
-
Tespl, Tessp5
- Chromosome Location
-
Chr.9: 110834588 - 110841310 on GRCm38
- UniGene ID
- Mm.79114
- Species
-
Mouse