gRNA Design Details
- Target DNA Sequence
- GCAACACATTACCTGGTGAA
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 127889577 - 127889599 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
85.95/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Prss53
- Entrez Gene ID
-
330657
- Gene Name
- protease, serine 53
- Gene Aliases
-
Pol3s, mFLJ00289
- Chromosome Location
-
Chr.7: 127885444 - 127890970 on GRCm38
- UniGene ID
- Mm.425491
- Species
-
Mouse