gRNA Design Details
- Target DNA Sequence
- GCTTCTTGGTGGACGCATCC
- PAM Sequence
- TGG
- Target locus
-
Chr.7: 45466035 - 45466057 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.01/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Bax
- Entrez Gene ID
-
12028
- Gene Name
- BCL2-associated X protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 45461695 - 45466903 on GRCm38
- UniGene ID
- Mm.19904
- Species
-
Mouse