gRNA Design Details
- Target DNA Sequence
- CTGCAACTTAATTGACAACG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 30549057 - 30549079 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.09/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Proser3
- Entrez Gene ID
-
333193
- Gene Name
- proline and serine rich 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 30539129 - 30552297 on GRCm38
- UniGene ID
- Mm.34885
- Species
-
Mouse