gRNA Design Details
- Target DNA Sequence
- TAGCCACCAAAAGTCAGCTG
- PAM Sequence
- AGG
- Target locus
-
Chr.7: 30546340 - 30546362 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
85.16/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Proser3
- Entrez Gene ID
-
333193
- Gene Name
- proline and serine rich 3
- Gene Aliases
-
-
- Chromosome Location
-
Chr.7: 30539129 - 30552297 on GRCm38
- UniGene ID
- Mm.34885
- Species
-
Mouse