gRNA Design Details
- Target DNA Sequence
- GGTCTTCCGGATCTTCAACC
- PAM Sequence
- CGG
- Target locus
-
Chr.6: 55472299 - 55472321 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
86.9/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Adcyap1r1
- Entrez Gene ID
-
11517
- Gene Name
- adenylate cyclase activating polypeptide 1 receptor 1
- Gene Aliases
-
2900024I10Rik, AI846590, PAC1, PAC1R, PACAP1-R
- Chromosome Location
-
Chr.6: 55451501 - 55501455 on GRCm38
- UniGene ID
- Mm.44245
- Species
-
Mouse