gRNA Design Details
- Target DNA Sequence
- GCCGCCACTTATGGGAAATG
- PAM Sequence
- TGG
- Target locus
-
Chr.13: 103761089 - 103761111 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
86.12/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srek1
- Entrez Gene ID
-
218543
- Gene Name
- splicing regulatory glutamine/lysine-rich protein 1
- Gene Aliases
-
8430401B01, AI450757, AI462342, SRrp508, SRrp86, Sfrs12, Srsf12
- Chromosome Location
-
Chr.13: 103739348 - 103774603 on GRCm38
- UniGene ID
- Mm.458176
- Species
-
Mouse