gRNA Design Details
- Target DNA Sequence
- ATTCGGAGGGCACCTCAGAC
- PAM Sequence
- AGG
- Target locus
-
Chr.10: 87913645 - 87913667 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igf1
- Entrez Gene ID
-
16000
- Gene Name
- insulin-like growth factor 1
- Gene Aliases
-
C730016P09Rik, Igf-1, Igf-I
- Chromosome Location
-
Chr.10: 87858270 - 87937047 on GRCm38
- UniGene ID
- Mm.268521
- Species
-
Mouse