gRNA Design Details
- Target DNA Sequence
- TTGGCCCCGGATCTTGGATA
- PAM Sequence
- CGG
- Target locus
-
Chr.17: 56019407 - 56019429 on GRCm38
- Strand
- Forward
- Application
- Gene Knockout
Score:
90.45/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sh3gl1
- Entrez Gene ID
-
20405
- Gene Name
- SH3-domain GRB2-like 1
- Gene Aliases
-
C77078, EEN, SH3P8, Sh3d2b
- Chromosome Location
-
Chr.17: 56016750 - 56036638 on GRCm38
- UniGene ID
- Mm.1773
- Species
-
Mouse