gRNA Design Details
- Target DNA Sequence
- GGACTCACCTGCATCTAGCA
- PAM Sequence
- GGG
- Target locus
-
Chr.17: 56019145 - 56019167 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
84.89/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Sh3gl1
- Entrez Gene ID
-
20405
- Gene Name
- SH3-domain GRB2-like 1
- Gene Aliases
-
C77078, EEN, SH3P8, Sh3d2b
- Chromosome Location
-
Chr.17: 56016750 - 56036638 on GRCm38
- UniGene ID
- Mm.1773
- Species
-
Mouse