gRNA Design Details

Target DNA Sequence
TTTCAACGTGAAGAAGCTGG
PAM Sequence
CGG
Target locus
Chr.3: 144720017 - 144720039 on GRCm38
Strand
Reverse
Application
Gene Knock-Out
Score: 84.91/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Sh3glb1
Entrez Gene ID
54673
Gene Name
SH3-domain GRB2-like B1 (endophilin)
Gene Aliases
AA409932, AI314629, AU015566, Bif-1, mKIAA0491
Chromosome Location
Chr.3: 144683678 - 144720398 on GRCm38
UniGene ID
Mm.271775
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001282037.1 NP_001268966.1 1
NM_001282042.1 NP_001268971.1 1
NM_019464.3 NP_062337.1 1