gRNA Design Details
- Target DNA Sequence
- GACTCGGGGTAGAACTTTCC
- PAM Sequence
- CGG
- Target locus
-
Chr.9: 110729636 - 110729658 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.69/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Pth1r
- Entrez Gene ID
-
19228
- Gene Name
- parathyroid hormone 1 receptor
- Gene Aliases
-
PPR, Pthr, Pthr1
- Chromosome Location
-
Chr.9: 110722085 - 110747145 on GRCm38
- UniGene ID
- Mm.3542
- Species
-
Mouse