gRNA Design Details
- Target DNA Sequence
- TATCTCCTTGTTTCACGTGC
- PAM Sequence
- TGG
- Target locus
-
Chr.14: 44989156 - 44989178 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
87.91/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ptger2
- Entrez Gene ID
-
19217
- Gene Name
- prostaglandin E receptor 2 (subtype EP2)
- Gene Aliases
-
EP2, Ptgerep2
- Chromosome Location
-
Chr.14: 44988111 - 45003820 on GRCm38
- UniGene ID
- Mm.4630
- Species
-
Mouse