gRNA Design Details
- Target DNA Sequence
- CTATCGATGCTCCATAGGCT
- PAM Sequence
- TGG
- Target locus
-
Chr.11: 74911066 - 74911088 on GRCm38
- Strand
- Forward
- Application
- Gene Knock-Out
Score:
86.97/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Srr
- Entrez Gene ID
-
27364
- Gene Name
- serine racemase
- Gene Aliases
-
Srs
- Chromosome Location
-
Chr.11: 74905047 - 74925798 on GRCm38
- UniGene ID
- Mm.131443
- Species
-
Mouse