gRNA Design Details
- Target DNA Sequence
- GCGAAAAGCGAATGATAAAC
- PAM Sequence
- AGG
- Target locus
-
Chr.8: 71678685 - 71678707 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
85.3/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Jak3
- Entrez Gene ID
-
16453
- Gene Name
- Janus kinase 3
- Gene Aliases
-
fae, wil
- Chromosome Location
-
Chr.8: 71676383 - 71690577 on GRCm38
- UniGene ID
- Mm.249645
- Species
-
Mouse