gRNA Design Details

Target DNA Sequence
TGAGTTGATAGGGCACCACT
PAM Sequence
TGG
Target locus
Chr.8: 83734097 - 83734119 on GRCm38
Strand
Forward
Application
Gene Knock-Out
Score: 84.54/100

Rank:
scoring gRNA

Targeted Gene

Gene Symbol
Adgre5
Entrez Gene ID
26364
Gene Name
adhesion G protein-coupled receptor E5
Gene Aliases
AA409984, Cd97, TM7LN1
Chromosome Location
Chr.8: 83723250 - 83741311 on GRCm38
UniGene ID
Mm.334648
Species
Mouse

Targeted Isoforms Translated Protein Targeted Exon(s)
RefSeq NM_001163029.1 NP_001156501.1 3
NM_001163030.1 NP_001156502.1 3
NM_001163031.1 NP_001156503.1 3
NM_011925.2 NP_036055.2 3