gRNA Design Details
- Target DNA Sequence
- ACGGCCAAGAAACAACAGTT
- PAM Sequence
- AGG
- Target locus
-
Chr.11: 7197970 - 7197992 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
84.04/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Igfbp1
- Entrez Gene ID
-
16006
- Gene Name
- insulin-like growth factor binding protein 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.11: 7197787 - 7202546 on GRCm38
- UniGene ID
- Mm.21300
- Species
-
Mouse