gRNA Design Details
- Target DNA Sequence
- GGAACAGGTCGGACATCACC
- PAM Sequence
- AGG
- Target locus
-
Chr.17: 29098417 - 29098439 on GRCm38
- Strand
- Reverse
- Application
- Gene Knock-Out
Score:
86.74/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Cdkn1a
- Entrez Gene ID
-
12575
- Gene Name
- cyclin-dependent kinase inhibitor 1A (P21)
- Gene Aliases
-
CAP20, CDKI, CIP1, Cdkn1, P21, SDI1, Waf1, mda6, p21Cip1, p21WAF
- Chromosome Location
-
Chr.17: 29090986 - 29100722 on GRCm38
- UniGene ID
- Mm.195663
- Species
-
Mouse