gRNA Design Details
- Target DNA Sequence
- GATCAACCTCACCTACAGGG
- PAM Sequence
- CGG
- Target locus
-
Chr.4: 88522609 - 88522631 on GRCm38
- Strand
- Reverse
- Application
- Gene Knockout
Score:
91.98/100
Rank:
scoring gRNA
Targeted Gene
- Gene Symbol
- Ifnb1
- Entrez Gene ID
-
15977
- Gene Name
- interferon beta 1, fibroblast
- Gene Aliases
-
IFN-beta, IFNB, Ifb
- Chromosome Location
-
Chr.4: 88522025 - 88522794 on GRCm38
- UniGene ID
- Mm.1245
- Species
-
Mouse